Assay Details
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159502.1 | NP_001152974.1 | 5 - 6 | 2172 | 80 | |
NM_020494.3 | NP_065240.2 | 5 - 6 | 2006 | 80 | ||
GenBank mRNA | AF214732.1 | - | 5 - 6 | 1986 | 80 | |
AK149810.1 | - | 5 - 6 | 1959 | 80 | ||
AK150778.1 | - | 5 - 6 | 1958 | 80 | ||
AK152551.1 | - | 5 - 6 | 1958 | 80 | ||
AK152928.1 | - | 5 - 6 | 1958 | 80 | ||
AK166360.1 | - | 5 - 6 | 1967 | 80 | ||
AK172557.1 | - | 5 - 6 | 1959 | 80 | ||
BC043721.1 | - | 5 - 6 | 1974 | 80 | ||
BC055048.1 | - | 5 - 6 | 1936 | 6484820 | 80 | |
BC055317.1 | - | 5 - 6 | 2172 | 5696084 | 80 | |
U46690.1 | - | 3 - 4 | 720 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
