Assay Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020603.2 | NP_065628.1 | 6 - 7 | 634 | 62 | |
| GenBank mRNA | AK216977.1 | - | 4 - 5 | 295 | 62 | |
| BC046977.1 | - | 6 - 7 | 634 | 6315172 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151139 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 37909 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161649 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161298 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31151152 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03309687 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03309676 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161298 | - | - |
| Whole Mouse Genome G4122A | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295313 | BC046977.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295313 | AK009209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295313 | NM_020603.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111853 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13156173 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13156158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31151139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31151133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13304224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13156161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13252869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13158767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13252873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13115891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13304229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13156164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13252870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13151288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13288778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31151140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444103 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437828_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422709_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444098 | - | - |
| Mouse Genome 430 2.0 Array | 1422709_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422709_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437828_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1437828_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57315_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57315_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700005625.mm.1 | NM_020603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005625.mm.1 | NM_020603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005625.mm.1 | TC1700000603.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700005624.mm.1 | NM_020603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005624.mm.1 | NM_020603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700005624.mm.1 | TC1700000603.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422709_a_at | - | - |
| Mouse Expression Set 430 | 1422709_PM_a_at | - | - |
| Murine Genome U74v2 Set | 104702_at | - | - |
| Murine Genome U74v2 Set | 104702_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422709_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422709_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422709_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422709_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437828_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437828_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 820967 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656889 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2656889 | - | - |
| mouse-6 v1.1 (Illumina) | scl50839.12.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50839.12.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2656894 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256643 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256643 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2656894 | - | - |

Wdr46
Genomic Map