Assay Details
| Gene Symbol: Ccdc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025602.3 | NP_079878.1 | 3 - 4 | 586 | 62 | |
| GenBank mRNA | AK012917.1 | - | 3 - 4 | 569 | 62 | |
| AK019970.1 | - | 3 - 4 | 587 | 62 | ||
| AK135550.1 | - | 3 - 4 | 611 | 62 | ||
| BC027401.1 | - | 3 - 4 | 568 | 4948072 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418183 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418184 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P129795 | AK135550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P129795 | BC027401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P129795 | NM_025602.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P129795 | AK012917.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P129795 | AK019970.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47943 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106375 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100076 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174746 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P129795 | AK135550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P129795 | BC027401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P129795 | NM_025602.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P129795 | AK012917.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P129795 | AK019970.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27418216 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106375 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174746 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100076 | - | - |
| Whole Mouse Genome G4122A | A_52_P557995 | AK047009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P557995 | AK135550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P557995 | BC013721.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P557995 | BC027401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P557995 | NM_025602.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129795 | AK135550.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129795 | BC027401.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129795 | NM_025602.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129795 | AK012917.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129795 | AK019970.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130360 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106375 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846709 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846708 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08846722 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418183 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27418184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27418278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08465997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08328310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08846723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08328311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08846711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08328303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27418199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08328313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10366189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366191 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449446_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10366186 | - | - |
| Mouse Genome 430 2.0 Array | 1449446_at | - | - |
| Mouse Genome 430 2.0 Array | 1449446_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52713_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52713_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000009445.mm.1 | NM_025602.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009445.mm.1 | NM_025602.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009445.mm.1 | TC1000001255.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000009447.mm.1 | NM_025602.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009447.mm.1 | NM_025602.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009447.mm.1 | TC1000001255.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000009446.mm.1 | NM_025602.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009446.mm.1 | NM_025602.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000009446.mm.1 | TC1000001255.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449446_at | - | - |
| Mouse Expression Set 430 | 1449446_PM_at | - | - |
| Murine Genome U74v2 Set | 167437_r_at | - | - |
| Murine Genome U74v2 Set | 167437_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449446_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449446_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449446_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449446_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 380181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4210273 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004210273 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380181 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712584 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256220 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256220 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712584 | - | - |
| mouse-6 v1.1 (Illumina) | scl0052713.1_274-S | - | - |
| mouse-6 v1.1 (Illumina) | scl052713.2_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380181 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4210273 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0052713.1_274-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl052713.2_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4210273 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256220 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2877436 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2712584 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2712584 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2877436 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256220 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl052713.2_45-S | - | - |

Ccdc59
Genomic Map