| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500001470.mm.1 |
NM_001161800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001470.mm.1 |
NM_001161800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001470.mm.1 |
NM_026448.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001470.mm.1 |
NM_026448.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001470.mm.1 |
TC0500000201.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001471.mm.1 |
NM_001161800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001471.mm.1 |
NM_001161800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001471.mm.1 |
NM_026448.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001471.mm.1 |
NM_026448.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001471.mm.1 |
TC0500000201.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500001472.mm.1 |
NM_001161800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001472.mm.1 |
NM_001161800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001472.mm.1 |
NM_026448.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001472.mm.1 |
NM_026448.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500001472.mm.1 |
TC0500000201.mm.1 |
MTA TC |