Assay Details
| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161800.1 | NP_001155272.1 | 3 - 4 | 475 | 122 | |
| NM_026448.3 | NP_080724.2 | 3 - 4 | 475 | 122 | ||
| GenBank mRNA | AK012326.1 | - | 3 - 4 | 456 | 122 | |
| AK082520.1 | - | 3 - 4 | 418 | 122 | ||
| AK083358.1 | - | 3 - 4 | 461 | 122 | ||
| AK089072.1 | - | 3 - 4 | 460 | 122 | ||
| BC029154.1 | - | 3 - 4 | 442 | 4952319 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl7
Genomic Map