Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 5 - 6 | 757 | 80 | |
| GenBank mRNA | AK012128.1 | - | 4 - 5 | 712 | 80 | |
| AK160356.1 | - | 5 - 6 | 726 | 80 | ||
| AK172381.1 | - | 4 - 5 | 500 | 80 | ||
| AK213160.1 | - | 3 - 4 | 246 | 80 | ||
| BC017625.1 | - | 4 - 5 | 706 | 80 | ||
More Information
Set Membership: |
|

Mpnd
Genomic Map