Assay Details
| Gene Symbol: Fam173b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026546.3 | NP_080822.1 | 2 - 3 | 314 | 121 | |
| GenBank mRNA | AK020868.1 | - | 2 - 3 | 314 | 121 | |
| AK160923.1 | - | 440 | 121 | |||
| BC030179.1 | - | 1 - 2 | 305 | 5039149 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115250 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52477 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154556 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161592 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30115259 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154556 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08326072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161592 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08326063 | - | - |
| Whole Mouse Genome G4122A | A_52_P502424 | AK044518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK160923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | BC030179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK020868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | AK044518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P117966 | NM_026546.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112781 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120340 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154556 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161592 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08326072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12140218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12140217 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30115250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30115371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12451714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12062763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11855650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12035980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11892086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12141399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12140218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12131378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12008816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12070105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12056207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11892087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12070107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12399986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12358964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11912140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30115277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12166840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12008817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12233129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11916538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11855649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12008809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10423515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423514 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444686_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428683_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10423512 | - | - |
| Mouse Genome 430 2.0 Array | 1428683_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444686_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457848_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457848_at | - | - |
| Mouse Genome 430 2.0 Array | 1428683_at | - | - |
| Mouse Genome 430 2.0 Array | 1444686_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68073_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68073_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500001544.mm.1 | NM_026546.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001544.mm.1 | NM_026546.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001544.mm.1 | TC1500000208.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444686_PM_at | - | - |
| Mouse Expression Set 430 | 1444686_at | - | - |
| Mouse Expression Set 430 | 1428683_PM_at | - | - |
| Mouse Expression Set 430 | 1428683_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444686_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428683_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428683_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444686_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 886949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105290520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105290520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 670546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224980 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224980 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2500032 | - | - |
| mouse-6 v1.1 (Illumina) | scl48019.6_564-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0068073.2_190-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105290520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068073.2_190-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670546 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105290520 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1224980 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224980 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0068073.2_190-S | - | - |

Fam173b
Genomic Map