Assay Details
| Gene Symbol: Cpsf4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164532.1 | NP_001158004.1 | 3 - 4 | 504 | 69 | |
| NM_026682.2 | NP_080958.2 | 3 - 4 | 345 | 69 | ||
| NM_029794.2 | NP_084070.1 | 3 - 4 | 401 | 69 | ||
| GenBank mRNA | AK005072.1 | - | 3 - 4 | 401 | 69 | |
| AK161074.1 | - | 3 - 4 | 346 | 69 | ||
| BC016190.1 | - | 3 - 4 | 339 | 4483305 | 69 | |
| DV648079.1 | - | 3 - 4 | 475 | 69 | ||
| DV659633.1 | - | 3 - 4 | 335 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf4l
Genomic Map