Assay Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032400.2 | NP_115776.2 | 1 - 2 | 52 | 83 | |
| GenBank mRNA | AF295367.1 | - | 1 - 2 | 68 | 83 | |
| AK080866.1 | - | 1 - 2 | 63 | 83 | ||
| BB864882.1 | - | 1 - 2 | 56 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF295367.1,NM_032400.2 transcripts.
Set Membership: |
|

Sucnr1
Genomic Map