Assay Details
| Gene Symbol: Il1rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133193.3 | NP_573456.1 | 1 - 2 | 195 | 74 | |
| GenBank mRNA | AF284433.1 | - | 1 - 2 | 200 | 74 | |
| BC117797.2 | - | 1 - 2 | 196 | 40092208 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF284433.1,BC117797.2,NM_133193.3 transcripts.
Set Membership: |
|

Il1rl2
Genomic Map