| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700008425.mm.1 |
NM_145353.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008425.mm.1 |
NM_145353.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008425.mm.1 |
TC1700000884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700008423.mm.1 |
NM_145353.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008423.mm.1 |
NM_145353.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008423.mm.1 |
TC1700000884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700008426.mm.1 |
NM_145353.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008426.mm.1 |
NM_145353.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008426.mm.1 |
TC1700000884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700008427.mm.1 |
NM_145353.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008427.mm.1 |
NM_145353.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700008427.mm.1 |
TC1700000884.mm.1 |
MTA TC |