Assay Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145353.2 | NP_663328.2 | 4 - 5 | 600 | 78 | |
| GenBank mRNA | AF197931.1 | - | 3 - 4 | 360 | 78 | |
| AK011822.1 | - | 4 - 5 | 587 | 78 | ||
| AK042644.1 | - | 3 - 4 | 272 | 78 | ||
| AK149957.1 | - | 4 - 5 | 433 | 78 | ||
| BC011161.1 | - | 4 - 5 | 561 | 78 | ||
| BC019384.1 | - | 4 - 5 | 581 | 5123736 | 78 | |
| BC100575.1 | - | 4 - 5 | 532 | 6432108 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yipf3
Genomic Map