Assay Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145353.2 | NP_663328.2 | 5 - 6 | 698 | 77 | |
| GenBank mRNA | AF197931.1 | - | 4 - 5 | 458 | 77 | |
| AK042644.1 | - | 4 - 5 | 370 | 77 | ||
| AK149957.1 | - | 5 - 6 | 531 | 77 | ||
| BC011161.1 | - | 5 - 6 | 659 | 77 | ||
| BC019384.1 | - | 5 - 6 | 679 | 5123736 | 77 | |
| BC100575.1 | - | 5 - 6 | 630 | 6432108 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yipf3
Genomic Map