Assay Details
| Gene Symbol: Yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145353.2 | NP_663328.2 | 6 - 7 | 827 | 63 | |
| GenBank mRNA | AK011822.1 | - | 6 - 7 | 814 | 63 | |
| AK042644.1 | - | 5 - 6 | 499 | 63 | ||
| AK149957.1 | - | 6 - 7 | 660 | 63 | ||
| BC011161.1 | - | 6 - 7 | 788 | 63 | ||
| BC019384.1 | - | 6 - 7 | 808 | 5123736 | 63 | |
| BC100575.1 | - | 6 - 7 | 759 | 6432108 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yipf3
Genomic Map