Assay Details
| Gene Symbol: Wfdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145369.3 | NP_663344.2 | 2 - 3 | 316 | 73 | |
| GenBank mRNA | AK131905.1 | - | 2 - 3 | 264 | 73 | |
| AK131906.1 | - | 2 - 3 | 249 | 73 | ||
| AY669505.1 | - | 2 - 3 | 315 | 73 | ||
| BC019734.1 | - | 2 - 3 | 316 | 3987617 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wfdc5
Genomic Map