Assay Details
| Gene Symbol: Wfdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145369.3 | NP_663344.2 | 3 - 4 | 483 | 73 | |
| GenBank mRNA | AK131905.1 | - | 3 - 4 | 431 | 73 | |
| AK131906.1 | - | 3 - 4 | 416 | 73 | ||
| AY669505.1 | - | 3 - 4 | 482 | 73 | ||
| BC019734.1 | - | 3 - 4 | 483 | 3987617 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098925 | AY669505.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098925 | NM_145369.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098925 | AK131905.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703466 | AK019857.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703466 | AK173252.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703466 | BC031499.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703466 | BC039211.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703466 | NM_026599.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137506 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138130 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098925 | AY669505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098925 | NM_145369.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098925 | AK131905.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21816683 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138130 | - | - |
| Whole Mouse Genome G4122A | A_51_P190509 | AY669505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190509 | BC019734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190509 | NM_145369.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190509 | AK075896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190509 | AK131905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190516 | AY669505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190516 | BC019734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190516 | NM_145369.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190516 | AK075896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190516 | AK131905.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133214 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138130 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02268002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01881294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02188241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01881295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02188236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21816658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02223339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02110586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01776945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01469278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02054692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02223336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01881294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01710251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01881295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21816676 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489432 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425418_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489429 | - | - |
| Mouse Genome 430 2.0 Array | 1425418_at | - | - |
| Mouse Genome 430 2.0 Array | 1425418_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209232_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209232_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043535.mm.1 | NM_145369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043535.mm.1 | NM_145369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043535.mm.1 | TC0200005105.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043534.mm.1 | NM_145369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043534.mm.1 | NM_145369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043534.mm.1 | TC0200005105.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043532.mm.1 | NM_145369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043532.mm.1 | NM_145369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043532.mm.1 | TC0200005105.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425418_PM_at | - | - |
| Mouse Expression Set 430 | 1425418_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425418_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425418_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425418_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425418_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 783617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6620609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225607 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225607 | - | - |
| mouse-6 v1.1 (Illumina) | scl18372.4.1_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18372.4.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225607 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225607 | - | - |

Wfdc5
Genomic Map