Assay Details
| Gene Symbol: Prmt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145404.1 | NP_663379.1 | 15 - 16 | 1781 | 78 | |
| GenBank mRNA | AK154255.1 | - | 15 - 16 | 1808 | 78 | |
| AK162376.1 | - | 375 | 78 | |||
| AK173304.1 | - | 15 - 16 | 1801 | 78 | ||
| AY673972.1 | - | 14 - 15 | 1651 | 78 | ||
| BC006705.1 | - | 15 - 16 | 1781 | 3583763 | 78 | |
| BC057177.1 | - | 15 - 16 | 1805 | 5010547 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt7
Genomic Map