Assay Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145417.3 | NP_663392.2 | 3 - 4 | 817 | 75 | |
| GenBank mRNA | AK145466.1 | - | 3 - 4 | 820 | 75 | |
| AK159195.1 | - | 3 - 4 | 760 | 75 | ||
| AK159445.1 | - | 3 - 4 | 749 | 75 | ||
| AK159625.1 | - | 3 - 4 | 760 | 75 | ||
| BC019200.1 | - | 3 - 4 | 788 | 5041005 | 75 | |
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK157889.1 | - | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rnpep
Genomic Map