Assay Details
Gene Symbol: Grhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161406.1 | NP_001154878.1 | 12 - 13 | 1651 | 62 | |
NM_145890.2 | NP_665897.2 | 10 - 11 | 1283 | 62 | ||
GenBank mRNA | AF411211.1 | - | 10 - 11 | 1283 | 62 | |
AF411212.1 | - | 12 - 13 | 1622 | 62 | ||
AK076307.1 | - | 12 - 13 | 1637 | 62 | ||
AK161048.1 | - | 12 - 13 | 1533 | 62 | ||
BC013080.1 | - | 12 - 13 | 1591 | 3499860 | 62 | |
BC037233.1 | - | 12 - 13 | 1643 | 4951512 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
