| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600012391.mm.1 |
NM_145932.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012391.mm.1 |
NM_145932.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012391.mm.1 |
TC1600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012387.mm.1 |
NM_145932.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012387.mm.1 |
NM_145932.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012387.mm.1 |
TC1600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012389.mm.1 |
NM_145932.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012389.mm.1 |
NM_145932.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012389.mm.1 |
TC1600001509.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600012388.mm.1 |
NM_145932.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012388.mm.1 |
NM_145932.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600012388.mm.1 |
TC1600001509.mm.1 |
MTA TC |