Assay Details
| Gene Symbol: Ly6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010742.1 | NP_034872.1 | 2 - 3 | 160 | 64 | |
| GenBank mRNA | BC025135.1 | - | 2 - 3 | 155 | 4914126 | 64 |
| X63782.1 | - | 2 - 3 | 160 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593920 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179486 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118753 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108878 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | X63782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339478 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179486 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108878 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118753 | - | - |
| Whole Mouse Genome G4122A | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P343517 | NM_010742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P343517 | BC025135.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P343517 | X63782.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106499 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108878 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179486 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095891 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11855915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12095897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11855918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11918538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12414585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11855905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12120630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12414578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12120623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11906594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12414574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12095903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12120624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12095901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12286073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12095891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11918536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12095894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12365124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12365125 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429522 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416930_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429520 | - | - |
| Mouse Genome 430 2.0 Array | 1416930_at | - | - |
| Mouse Genome 430 2.0 Array | 1416930_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17068_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17068_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500012971.mm.1 | NM_010742.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012971.mm.1 | NM_010742.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012971.mm.1 | TC1500001718.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416930_at | - | - |
| Mouse Expression Set 430 | 1416930_PM_at | - | - |
| Murine Genome U74v2 Set | 160553_at | - | - |
| Murine Genome U74v2 Set | 160553_PM_at | - | - |
| Murine Genome U74v2 Set | 162216_i_at | - | - |
| Murine Genome U74v2 Set | 162216_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416930_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416930_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416930_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416930_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 577604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4050010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257839 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257839 | - | - |
| mouse-6 v1.1 (Illumina) | scl47080.2.4_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47080.2.4_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803920 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803921 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803920 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803921 | - | - |

Ly6d
Genomic Map