Assay Details
| Gene Symbol: Magea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020016.1 | NP_064400.1 | 1 - 2 | 41 | 72 | |
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0X00003231.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0X00003231.mm.1Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the NM_020016.1 transcripts.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Ambion siRNA available  Amplicon length between 71 and 85  Mus musculus  Inventoried  Probe spans exons | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Magea2
Magea2 Genomic Map
Genomic Map Back To Top
 Back To Top