Assay Details
| Gene Symbol: Mark3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021516.4 | NP_067491.2 | 8 - 9 | 1314 | 58 | |
| NM_022801.4 | NP_073712.2 | 8 - 9 | 1314 | 58 | ||
| GenBank mRNA | AF240782.1 | - | 8 - 9 | 859 | 58 | |
| AF240783.1 | - | 8 - 9 | 859 | 58 | ||
| AK083683.1 | - | 8 - 9 | 1315 | 58 | ||
| AK132247.1 | - | 8 - 9 | 1298 | 58 | ||
| AK166157.1 | - | 8 - 9 | 1303 | 58 | ||
| AK220557.1 | - | 7 - 8 | 5148 | 58 | ||
| BC026445.1 | - | 1 - 2 | 219 | 4459439 | 58 | |
| BC066071.1 | - | 8 - 9 | 1288 | 58 | ||
| CX566304.1 | - | 5 - 6 | 452 | 58 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC026445.1 transcripts.
Set Membership: |
|

Mark3
Genomic Map