Assay Details
| Gene Symbol: Mark3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021516.4 | NP_067491.2 | 9 - 10 | 1438 | 63 | |
| NM_022801.4 | NP_073712.2 | 9 - 10 | 1438 | 63 | ||
| GenBank mRNA | AF240782.1 | - | 9 - 10 | 983 | 63 | |
| AF240783.1 | - | 9 - 10 | 983 | 63 | ||
| AK083683.1 | - | 9 - 10 | 1439 | 63 | ||
| AK132247.1 | - | 9 - 10 | 1422 | 63 | ||
| AK166157.1 | - | 9 - 10 | 1427 | 63 | ||
| AK217056.1 | - | 1 - 2 | 99 | 63 | ||
| AK220557.1 | - | 8 - 9 | 5272 | 63 | ||
| BC026445.1 | - | 2 - 3 | 343 | 4459439 | 63 | |
| BC066071.1 | - | 9 - 10 | 1412 | 63 | ||
| CX566304.1 | - | 6 - 7 | 576 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC026445.1 transcripts.
Set Membership: |
|

Mark3
Genomic Map