Assay Details
| Gene Symbol: Lgi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144556.2 | NP_653139.2 | 3 - 4 | 395 | 93 | |
| GenBank mRNA | AJ487521.1 | - | 3 - 4 | 396 | 93 | |
| AJ487960.1 | - | 3 - 4 | 395 | 93 | ||
| AK033967.1 | - | 3 - 4 | 213 | 93 | ||
| AK134643.1 | - | 3 - 4 | 989 | 93 | ||
| AK148793.1 | - | 3 - 4 | 654 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034847 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034849 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033312 | AJ487960.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033312 | AK134643.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033312 | NM_144556.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033312 | BC051033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033312 | EH101686.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47088 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125701 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149974 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033312 | AJ487960.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033312 | AK134643.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033312 | NM_144556.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033312 | BC051033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033312 | EH101686.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034807 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149974 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01494066 | - | - |
| Whole Mouse Genome G4122A | A_52_P374556 | BC098469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374556 | AJ487960.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374556 | AK134643.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374556 | NM_144556.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374556 | BC051033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217386 | BC098469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217386 | AJ487960.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217386 | AK033967.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217386 | AK134643.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217386 | NM_144556.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140145 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125701 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05952460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05952464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034848 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034849 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05952464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06283647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06198300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05925096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06438259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06438266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06283648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25034823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552081 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434121_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562223 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552075 | - | - |
| Mouse Genome 430 2.0 Array | 1434121_at | - | - |
| Mouse Genome 430 2.0 Array | 1434121_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243914_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243914_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700005242.mm.1 | NM_144556.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005242.mm.1 | NM_144556.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700005242.mm.1 | TC0700000586.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434121_PM_at | - | - |
| Mouse Expression Set 430 | 1434121_at | - | - |
| Murine Genome U74v2 Set | 103060_PM_at | - | - |
| Murine Genome U74v2 Set | 103060_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434121_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434121_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434121_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434121_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 559494 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1570577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001570577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639386 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2525815 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639386 | - | - |
| mouse-6 v1.1 (Illumina) | scl32801.8.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1570577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32801.8.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639386 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639386 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32801.8.1_2-S | - | - |

Lgi4
Genomic Map