Assay Details
| Gene Symbol: Ankrd33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144790.1 | NP_659039.1 | 1 - 2 | 151 | 77 | |
| GenBank mRNA | BC022726.1 | - | 1 - 2 | 151 | 4504539 | 77 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC022726.1,NM_144790.1 transcripts.
Set Membership: |
|

Ankrd33
Genomic Map