Assay Details
| Gene Symbol: Ankrd33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144790.1 | NP_659039.1 | 2 - 3 | 282 | 65 | |
| GenBank mRNA | AK044543.1 | - | 3 - 4 | 467 | 65 | |
| AK044546.1 | - | 2 - 3 | 260 | 65 | ||
| BC022726.1 | - | 2 - 3 | 282 | 4504539 | 65 | |
| FJ895380.1 | - | 3 - 4 | 443 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd33
Genomic Map