Assay Details
| Gene Symbol: Pycr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144795.3 | NP_659044.1 | 2 - 3 | 582 | 82 | |
| GenBank mRNA | AK139185.1 | - | 2 - 3 | 583 | 82 | |
| BC006727.1 | - | 3 - 4 | 227 | 3962093 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proline biosynthetic process
cellular amino acid biosynthetic process
cellular response to oxidative stress
regulation of mitochondrial membrane potential
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
pyrroline-5-carboxylate reductase activity
oxidoreductase activity
identical protein binding
proline biosynthetic process
cellular amino acid biosynthetic process
cellular response to oxidative stress
regulation of mitochondrial membrane potential
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
pyrroline-5-carboxylate reductase activity
oxidoreductase activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169234 | AK139185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169234 | NM_144795.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673925 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110592 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169234 | AK139185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169234 | NM_144795.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110592 | - | - |
| Whole Mouse Genome G4122A | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P503896 | BC006727.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107481 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110592 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09288455 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09288468 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08984308 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08984330 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08984342 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08984306 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09288459 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09288450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09642337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09652070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09337044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09116190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09455058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09148018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09337049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08984315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185885 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393893 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424556_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393887 | - | - |
| Mouse Genome 430 2.0 Array | 1424556_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424556_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100040382.mm.1 | NM_144795.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040382.mm.1 | NM_144795.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040382.mm.1 | TC1100004222.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040381.mm.1 | NM_144795.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040381.mm.1 | NM_144795.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040381.mm.1 | TC1100004222.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424556_PM_at | - | - |
| Mouse Expression Set 430 | 1424556_at | - | - |
| Murine Genome U74v2 Set | 165691_at | - | - |
| Murine Genome U74v2 Set | 165691_PM_at | - | - |
| Murine Genome U74v2 Set | 167490_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165692_PM_i_at | - | - |
| Murine Genome U74v2 Set | 165692_i_at | - | - |
| Murine Genome U74v2 Set | 167490_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424556_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424556_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424556_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424556_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 560198 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6180670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664155 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664155 | - | - |
| mouse-6 v1.1 (Illumina) | scl39227.8.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39227.8.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180670 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2849521 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664155 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2849521 | - | - |

Pycr1
Genomic Map