Assay Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144849.1 | NP_659098.1 | 3 - 4 | 535 | 59 | |
| GenBank mRNA | AK050440.1 | - | 3 - 4 | 562 | 59 | |
| AK133785.1 | - | 3 - 4 | 559 | 59 | ||
| AK143625.1 | - | 3 - 4 | 562 | 59 | ||
| BC014743.1 | - | 3 - 4 | 535 | 4503319 | 59 | |
| DQ884878.1 | - | 3 - 4 | 563 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd54
Genomic Map