| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_001144987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_001144987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_001144988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_001144988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_144895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
NM_144895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002940.mm.1 |
TC0300000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_001144987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_001144987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_001144988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_001144988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_144895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
NM_144895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002945.mm.1 |
TC0300000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_001144987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_001144987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_001144988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_001144988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_144895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
NM_144895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002946.mm.1 |
TC0300000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_001144987.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_001144987.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_001144988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_001144988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_144895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
NM_144895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002947.mm.1 |
TC0300000416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002939.mm.1 |
NM_001144988.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002939.mm.1 |
NM_001144988.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002939.mm.1 |
NM_144895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002939.mm.1 |
NM_144895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002939.mm.1 |
TC0300000416.mm.1 |
MTA TC |