Assay Details
| Gene Symbol: Tmem150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144916.3 | NP_659165.1 | 2 - 3 | 243 | 59 | |
| GenBank mRNA | BC014685.1 | - | 1 - 2 | 159 | 4241375 | 59 |
| BC028825.1 | - | 2 - 3 | 245 | 5097962 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem150a
Genomic Map