Assay Details
| Gene Symbol: Mettl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110512.1 | NP_001103982.1 | 1 - 2 | 154 | 104 | |
| NM_145524.3 | NP_663499.2 | 2 - 3 | 390 | 104 | ||
| GenBank mRNA | AK039461.1 | - | 2 - 3 | 357 | 104 | |
| AK042669.1 | - | 2 - 3 | 368 | 104 | ||
| BC004636.1 | - | 2 - 3 | 311 | 3585719 | 104 | |
| DQ093357.1 | - | 2 - 3 | 367 | 104 | ||
| DQ093358.1 | - | 2 - 3 | 367 | 104 | ||
| DQ230973.1 | - | 1 - 2 | 158 | 104 | ||
| DQ230974.1 | - | 1 - 2 | 158 | 104 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC004636.1 transcripts.
Set Membership: |
|

Mettl8
Genomic Map