Assay Details
| Gene Symbol: Mettl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110512.1 | NP_001103982.1 | 3 - 4 | 360 | 72 | |
| NM_145524.3 | NP_663499.2 | 4 - 5 | 824 | 72 | ||
| GenBank mRNA | AK039461.1 | - | 4 - 5 | 563 | 72 | |
| BC004636.1 | - | 4 - 5 | 745 | 3585719 | 72 | |
| BC057960.1 | - | 4 - 5 | 691 | 5354784 | 72 | |
| DQ093358.1 | - | 4 - 5 | 801 | 72 | ||
| DQ230973.1 | - | 3 - 4 | 592 | 72 | ||
| DQ230974.1 | - | 3 - 4 | 364 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl8
Genomic Map