Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145549.1 | NP_663524.1 | 860 | 108 | ||
| GenBank mRNA | AK142188.1 | - | 4 - 5 | 1065 | 108 | |
| AK143777.1 | - | 918 | 108 | |||
| AK220422.1 | - | 809 | 108 | |||
| BC020077.1 | - | 860 | 4018470 | 108 | ||
| BC112327.1 | - | 4 - 5 | 711 | 5362489 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab7
Genomic Map