Assay Details
| Gene Symbol: Ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145610.2 | NP_663585.2 | 5 - 6 | 639 | 59 | |
| GenBank mRNA | AK039027.1 | - | 5 - 6 | 544 | 59 | |
| BC014688.1 | - | 5 - 6 | 557 | 4489442 | 59 | |
| BC059277.1 | - | 4 - 5 | 723 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344350 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344349 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344351 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169510 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140288 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26344353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140288 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06717200 | - | - |
| Whole Mouse Genome G4122A | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | AK172615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | BC014688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | BC059277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189985 | AK171672.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122645 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140288 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08025787 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344350 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344349 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26344351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26344382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08025784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07955393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07955416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08025781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07976533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07955418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08047244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07770913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08022729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08022730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07955417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26344365 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583500 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423703_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583496 | - | - |
| Mouse Genome 430 2.0 Array | 1423703_at | - | - |
| Mouse Genome 430 2.0 Array | 1423703_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235036_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235036_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900001171.mm.1 | NM_145610.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001171.mm.1 | NM_145610.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900001171.mm.1 | TC0900000182.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423703_at | - | - |
| Mouse Expression Set 430 | 1423703_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423703_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423703_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423703_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423703_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 666365 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 540398 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 540398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000540398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2772174 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2772174 | - | - |
| mouse-6 v1.1 (Illumina) | scl37238.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 540398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37238.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2778530 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778530 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2772174 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37238.1_20-S | - | - |

Ppan
Genomic Map