Assay Details
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145610.2 | NP_663585.2 | 9 - 10 | 1029 | 94 | |
GenBank mRNA | AK039027.1 | - | 9 - 10 | 934 | 94 | |
AK171672.1 | - | 5 - 6 | 695 | 94 | ||
AK172615.1 | - | 5 - 6 | 695 | 94 | ||
BC014688.1 | - | 9 - 10 | 947 | 4489442 | 94 | |
BC059277.1 | - | 7 - 8 | 1193 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
