Assay Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145958.2 | NP_666070.2 | 4 | 2330 | 88 | |
| GenBank mRNA | AK129374.1 | - | 4 | 2240 | 88 | |
| BC022962.1 | - | 1 | 1247 | 5368323 | 88 | |
| BC023724.1 | - | 1 | 1119 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964852 | BC022962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964852 | AK129374.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964852 | NM_145958.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333798 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117212 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113249 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121202 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964852 | BC022962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964852 | AK129374.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964852 | NM_145958.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24476790 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05910436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121202 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113249 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01337406 | - | - |
| Whole Mouse Genome G4122A | A_52_P318518 | BC022962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318518 | AK129374.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P318518 | NM_145958.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189151 | AK028396.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129718 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113249 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121202 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05910436 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05856100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05856111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05082005 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05122762 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05122759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24476812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05039862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05209136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05233950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05299969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05336563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05100937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24476940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10544977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544978 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451214_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544976 | - | - |
| Mouse Genome 430 2.0 Array | 1451214_at | - | - |
| Mouse Genome 430 2.0 Array | 1451214_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210973_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210973_PM_at | - | - |
| Mouse Expression Set 430 | 1451214_at | - | - |
| Mouse Expression Set 430 | 1451214_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451214_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451214_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451214_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451214_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 614063 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105720040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105720040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3060452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003060452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2654442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2654442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254564 | - | - |
| mouse-6 v1.1 (Illumina) | scl28965.7_274-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3060452 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105720040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28965.7_274-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060452 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105720040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2824593 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2654442 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254564 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654442 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2824593 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28965.7_274-S | - | - |

Kbtbd2
Genomic Map