Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177798.1 | NP_001171269.1 | 10 - 11 | 1297 | 91 | |
| NM_146092.2 | NP_666204.2 | 10 - 11 | 1134 | 91 | ||
| GenBank mRNA | AK077149.1 | - | 10 - 11 | 1298 | 91 | |
| BC020299.1 | - | 10 - 11 | 1107 | 91 | ||
| BC028647.1 | - | 10 - 11 | 1081 | 1244243 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943530 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943528 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943532 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135383 | AK082962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135383 | AK085400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135383 | NR_015538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135383 | AK041085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135383 | AK077736.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783404 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111493 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164823 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135383 | AK082962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135383 | AK085400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135383 | NR_015538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135383 | AK041085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135383 | AK077736.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P231658 | AK082962.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P231658 | AK085400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P231658 | NR_015538.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P231658 | AK041085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P231658 | AK077736.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943543 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09100585 | - | - |
| Whole Mouse Genome G4122A | A_52_P231658 | AK082962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231658 | AK085400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231658 | NR_015538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231658 | AK041085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231658 | AK077736.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09100584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14491826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14491825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14384267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14384268 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14384243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14491823 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943528 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943532 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14285170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14285173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943533 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465801 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465802 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465803 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465799 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452600_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465790 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465783 | - | - |
| Mouse Genome 430 2.0 Array | 1452600_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452600_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225895_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225895_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900015575.mm.1 | NM_001177798.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015575.mm.1 | NM_001177798.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015575.mm.1 | NM_146092.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015575.mm.1 | NM_146092.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900015575.mm.1 | TC1900001770.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009989.mm.1 | NM_001177798.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009989.mm.1 | NM_001177798.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009989.mm.1 | NM_146092.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009989.mm.1 | NM_146092.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009989.mm.1 | TC1900001770.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009993.mm.1 | NM_001177798.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009993.mm.1 | NM_001177798.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009993.mm.1 | NM_146092.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009993.mm.1 | NM_146092.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009993.mm.1 | TC1900001770.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009990.mm.1 | NM_001177798.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009990.mm.1 | NM_001177798.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009990.mm.1 | NM_146092.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009990.mm.1 | NM_146092.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009990.mm.1 | TC1900001770.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009992.mm.1 | NM_001177798.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009992.mm.1 | NM_001177798.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009992.mm.1 | NM_146092.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009992.mm.1 | NM_146092.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009992.mm.1 | TC1900001770.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452600_PM_at | - | - |
| Mouse Expression Set 430 | 1452600_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452600_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452600_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452600_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452600_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1850056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850056 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594433 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594433 | - | - |
| mouse-6 v1.1 (Illumina) | scl00225895.1_99-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00225895.1_99-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594433 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594433 | - | - |

Taf6l
Genomic Map