Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC026926.1 | - | 1 - 2 | 159 | 3586097 | 79 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC026926.1 transcripts.
Set Membership: |
|

C87436
Genomic Map