Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243741.1 | NP_001230670.1 | 4 - 5 | 1777 | 59 | |
| NM_001243742.1 | NP_001230671.1 | 4 - 5 | 1343 | 59 | ||
| NM_146170.4 | NP_666282.3 | 4 - 5 | 1284 | 59 | ||
| GenBank mRNA | AK047127.1 | - | 4 - 5 | 1285 | 59 | |
| AK077101.1 | - | 1110 | 59 | |||
| AK077503.1 | - | 4 - 5 | 1664 | 59 | ||
| AK170524.1 | - | 5 - 6 | 1175 | 59 | ||
| AK171511.1 | - | 5 - 6 | 1175 | 59 | ||
| BC025632.1 | - | 4 - 5 | 1251 | 5321669 | 59 | |
| BC026926.1 | - | 4 - 5 | 1354 | 3586097 | 59 | |
| BC031913.1 | - | 4 - 5 | 1297 | 5376246 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

C87436
Genomic Map