Assay Details
Gene Symbol: Cnot3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146176.3 | NP_666288.1 | 3 - 4 | 461 | 90 | |
GenBank mRNA | AK141718.1 | - | 3 - 4 | 441 | 90 | |
BC030332.1 | - | 3 - 4 | 390 | 5400871 | 90 | |
BC053437.1 | - | 3 - 4 | 222 | 6394085 | 90 | |
BC141041.1 | - | 3 - 4 | 261 | 9055653 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
