Assay Details
Gene Symbol: Cnot3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146176.3 | NP_666288.1 | 8 - 9 | 1075 | 116 | |
GenBank mRNA | AK141718.1 | - | 8 - 9 | 1055 | 116 | |
AK173003.1 | - | 4 - 5 | 489 | 116 | ||
BC030332.1 | - | 8 - 9 | 1004 | 5400871 | 116 | |
BC053437.1 | - | 8 - 9 | 836 | 6394085 | 116 | |
BC141041.1 | - | 8 - 9 | 875 | 9055653 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
