Assay Details
| Gene Symbol: Olfr780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146284.1 | NP_666396.1 | 1 | 444 | 89 | |
| GenBank mRNA | BC138405.1 | - | 1838 | 8861425 | 89 | |
| BC138406.1 | - | 1838 | 8861426 | 89 | ||
| BC147436.1 | - | 1 | 500 | 9087435 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726714 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179932 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545461 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179932 | - | - |
| Whole Mouse Genome G4122A | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | BC138406.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | BC147436.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | NM_146284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P477229 | BC138405.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131216 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110537 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179932 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688695 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688690 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688693 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688689 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08688696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08254420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08627883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08468705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08688698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08468706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545470 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367486 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367485 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000012028.mm.1 | TC1000001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000012030.mm.1 | TC1000001637.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000012029.mm.1 | NM_146284.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000012029.mm.1 | NM_146284.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000012029.mm.1 | TC1000001637.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 767432 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900008 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240002 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240002 | - | - |
| mouse-6 v1.1 (Illumina) | scl38262.1.1_214-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38262.1.1_214-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240002 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240002 | - | - |

Olfr780
Genomic Map