Assay Details
| Gene Symbol: Olfr574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146360.2 | NP_666472.2 | 1 | 351 | 109 | |
| GenBank mRNA | BC127050.1 | - | 1 | 349 | 40081239 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152191 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25381353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152191 | - | - |
| Whole Mouse Genome G4122A | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P444822 | BC127050.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108162 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119997 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237667 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237660 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25381388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25381341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555755 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555753 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012707.mm.1 | NM_146360.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012707.mm.1 | NM_146360.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012707.mm.1 | TC0700001472.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 437767 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6290687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290687 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2774654 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2774654 | - | - |
| mouse-6 v1.1 (Illumina) | scl9405.1.1_291-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290687 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl9405.1.1_291-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2774654 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2774654 | - | - |

Olfr574
Genomic Map