Assay Details
| Gene Symbol: Olfr1451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146705.1 | NP_666916.1 | 1 | 682 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989399 | NM_146705.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108992 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989399 | NM_146705.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P347945 | NM_146705.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31963275 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108992 | - | - |
| Whole Mouse Genome G4122A | A_51_P347945 | NM_146705.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108916 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108992 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14209885 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14209886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14209879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14512382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31963299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14353137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14209886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14209885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31963299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461789 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 595075 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254141 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254141 | - | - |
| mouse-6 v1.1 (Illumina) | scl6291.1.1_113-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6291.1.1_113-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254141 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254141 | - | - |

Olfr1451
Genomic Map