Assay Details
| Gene Symbol: Olfr1104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146767.2 | NP_666978.2 | 1 | 851 | 128 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947773 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162894 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162894 | - | - |
| Whole Mouse Genome G4122A | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485854 | NM_146767.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137000 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21404527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01517955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01843133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01664313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02176905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01517956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21404518 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484649 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 427416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4810279 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810279 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2673611 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2673611 | - | - |
| mouse-6 v1.1 (Illumina) | scl4322.1.1_89-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810279 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4322.1.1_89-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810279 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2673610 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2673610 | - | - |

Olfr1104
Genomic Map