Assay Details
| Gene Symbol: Olfr1395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146877.1 | NP_667088.1 | 1 | 48 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781503 | - | - |
| Whole Mouse Genome G4122A | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P264866 | NM_146877.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117175 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09528251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09137238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09524827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09524823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09486719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09036495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09524827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09420902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09009700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09009694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09422975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09524823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375526 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375525 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 617847 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4230348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2623376 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2623376 | - | - |
| mouse-6 v1.1 (Illumina) | scl14142.1.1_88-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14142.1.1_88-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2623376 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2623376 | - | - |

Olfr1395
Genomic Map