Assay Details
| Gene Symbol: Olfr851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146905.2 | NP_667116.2 | 1 | 243 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952970 | NM_146905.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050707 | NM_146905.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124051 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050707 | NM_146905.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952970 | NM_146905.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218789 | NM_146905.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26340110 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124051 | - | - |
| Whole Mouse Genome G4122A | A_51_P218789 | NM_146905.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122696 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07542467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08043251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07734840 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08043254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07542432 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08043260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26340093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08107460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07734840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07542461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08043252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26340143 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583399 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583398 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900000978.mm.1 | NM_146905.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000978.mm.1 | NM_146905.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000978.mm.1 | TC0900000151.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 404914 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 630364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622050 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622050 | - | - |
| mouse-6 v1.1 (Illumina) | scl37254.1.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37254.1.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2622050 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622050 | - | - |

Olfr851
Genomic Map