Assay Details
| Gene Symbol: Olfr1260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146981.1 | NP_667192.1 | 1 | 438 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21415352 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177151 | - | - |
| Whole Mouse Genome G4122A | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P378903 | NM_146981.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02120010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01215191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01215190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01862329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02120012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02120005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01668589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01862329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01215190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01322320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21415373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473631 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473630 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200010145.mm.1 | NM_146981.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010145.mm.1 | NM_146981.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010145.mm.1 | TC0200001339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200010147.mm.1 | NM_146981.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010147.mm.1 | NM_146981.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010147.mm.1 | TC0200001339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200010143.mm.1 | NM_146981.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010143.mm.1 | NM_146981.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010143.mm.1 | TC0200001339.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200010144.mm.1 | NM_146981.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010144.mm.1 | NM_146981.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200010144.mm.1 | TC0200001339.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 570616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1850563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2627705 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2627705 | - | - |
| mouse-6 v1.1 (Illumina) | scl4889.1.1_194-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4889.1.1_194-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2627705 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2627705 | - | - |

Olfr1260
Genomic Map