Assay Details
| Gene Symbol: Olfr641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147072.1 | NP_667283.1 | 1 | 451 | 56 | |
| GenBank mRNA | BC127981.1 | - | 1 | 455 | 40135228 | 56 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143063 | - | - |
| Whole Mouse Genome G4122A | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419503 | NM_147072.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140965 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25386340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05890218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06627108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05972935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06056016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06183046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25386307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555843 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555842 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012812.mm.1 | NM_147072.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012812.mm.1 | NM_147072.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012812.mm.1 | TC0700001516.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 355454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4150725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2703233 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2703233 | - | - |
| mouse-6 v1.1 (Illumina) | scl32260.1.1_189-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32260.1.1_189-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2703233 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2703233 | - | - |

Olfr641
Genomic Map